Gene Transcripts

Gene Symbol
Ngfr
Entrez Gene ID
24596
Gene Name
nerve growth factor receptor
Gene Aliases
LNGFR, RNNGFRR, Tnfrsf16, p75, p75NTR
Chromosome Location
Chr. 10: 83389835 - 83408061 on Build Rnor_6.0
UniGene ID
Rn.10980
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012610.2 NP_036742.2 3 433
GenBank mRNA CB725789.1 - 3 441
X05137.1 - 3 433

View Full Product Details