Gene Transcripts

Gene Symbol
Psip1
Entrez Gene ID
313323
Gene Name
PC4 and SFRS1 interacting protein 1
Gene Aliases
Ledgf, Psip2, SBP75
Chromosome Location
Chr. 5: 101556198 - 101588664 on Build Rnor_6.0
UniGene ID
Rn.93075
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175765.2 NP_786941.1 3, 4 396
XM_006238353.2 XP_006238415.1 4, 5 620
XM_006238354.2 XP_006238416.1 4, 5 619
XM_008763779.1 XP_008762001.1 4, 5 619
GenBank mRNA AB285525.1 - 299
AF339084.1 - 3, 4 396
AF372090.1 - 3, 4 396
BC093606.1 - 4, 5 554 7317952

View Full Product Details