Gene Transcripts

Gene Symbol
Naprt1
Entrez Gene ID
315085
Gene Name
nicotinate phosphoribosyltransferase domain containing 1
Gene Aliases
Naprt
Chromosome Location
Chr. 7: 116922979 - 116926436 on Build Rnor_6.0
UniGene ID
Rn.22665
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207609.1 NP_997492.1 13 1754
GenBank mRNA AY214330.1 - 13 1754

View Full Product Details