Gene Transcripts

Gene Symbol
SNX10
Entrez Gene ID
29887
Gene Name
sorting nexin 10
Gene Aliases
OPTB8
Chromosome Location
Chr. 7: 26291895 - 26374330 on Build GRCh38
UniGene ID
Hs.741316
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199835.1 NP_001186764.1 3 339
NM_001199837.1 NP_001186766.1 1 144
NM_001318198.1 NP_001305127.1 4 413
NM_001318199.1 NP_001305128.1 3 214
NM_013322.2 NP_037454.2 3 292
XM_006715712.2 XP_006715775.1 3 492
XM_017012085.1 XP_016867574.1 5 507
XM_017012086.1 XP_016867575.1 5 946
XM_017012087.1 XP_016867576.1 4 436
XM_017012088.1 XP_016867577.1 4 383
GenBank mRNA AF121860.1 - 3 205
AK296142.1 - 4 243
AK307905.1 - 1 93
AK312850.1 - 3 214
AK316088.1 - 4 389
BC031050.1 - 3 186 5278015
BC034992.1 - 3 265 4827599
BJ995940.1 - 1 144
BM147978.1 - 3 333
DA377368.1 - 3 214

View Full Product Details