Gene Transcripts

Gene Symbol
TMEM50A
Entrez Gene ID
23585
Gene Name
transmembrane protein 50A
Gene Aliases
IFNRC, SMP1
Chromosome Location
Chr. 1: 25338298 - 25362361 on Build GRCh38
UniGene ID
Hs.705699
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014313.3 NP_055128.1 7 981
XM_005245817.1 XP_005245874.1 6 852
XM_011541159.1 XP_011539461.1 7 1175
GenBank mRNA AF081282.1 - 7 909
AK299180.1 - 6 852
AK311092.1 - 4 659
AK311355.1 - 5 726
AL136627.1 - 7 963
AY071927.1 - 7 958
AY358650.1 - 7 944
BC066908.1 - 7 920 4817784
BX537683.1 - 1 3905
JX644905.1 - 7 981
JX644906.1 - 7 981

View Full Product Details