Gene Transcripts
- Gene Symbol
- PLPP2
- Entrez Gene ID
- 8612
- Gene Name
- phospholipid phosphatase 2
- Gene Aliases
- LPP2, PAP-2c, PAP2-g, PPAP2C
- Chromosome Location
- Chr. 19: 281040 - 291435 on Build GRCh38
- UniGene ID
- Hs.465506
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_003712.3 |
NP_003703.1
|
3, 4 |
584 |
|
NM_177526.2 |
NP_803545.1
|
3, 4 |
543 |
|
NM_177543.2 |
NP_808211.1
|
3, 4 |
651 |
|
XM_011528396.2 |
XP_011526698.1
|
3, 4 |
580 |
|
GenBank mRNA |
AF035959.1 |
- |
3, 4 |
528 |
|
AF047760.1 |
- |
3, 4 |
528 |
|
AF056083.1 |
- |
3, 4 |
528 |
|
BC002806.2 |
- |
3, 4 |
535 |
3659728 |
BM050291.1 |
- |
3, 4 |
585 |
|
BT007021.1 |
- |
3, 4 |
485 |
|
View Full Product Details