Gene Transcripts

Gene Symbol
HERC4
Entrez Gene ID
26091
Gene Name
HECT and RLD domain containing E3 ubiquitin protein ligase 4
Gene Aliases
Chromosome Location
Chr. 10: 67921899 - 68075348 on Build GRCh38
UniGene ID
Hs.51891
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278185.1 NP_001265114.1 22, 23 2935
NM_001278186.1 NP_001265115.1 21, 22 2893
NM_015601.3 NP_056416.2 23, 24 3145
NM_022079.2 NP_071362.1 24, 25 3169
XM_011539592.2 XP_011537894.1 25, 26 3243
XM_011539593.2 XP_011537895.1 24, 25 3219
XM_011539594.2 XP_011537896.1 26, 27 3329
XM_011539595.2 XP_011537897.1 26, 27 3379
XM_011539596.2 XP_011537898.1 25, 26 3257
XM_011539597.2 XP_011537899.1 23, 24 2880
XM_017016041.1 XP_016871530.1 22, 23 2808
GenBank mRNA AB046813.1 - 21, 22 3023
AK027835.1 - 14, 15 1708
AK292809.1 - 23, 24 3025
AY221963.1 - 24, 25 3106
AY650032.1 - 21, 22 2830
AY650033.1 - 22, 23 2872
BC015043.1 - 21, 22 2796
BC039600.1 - 23, 24 3055 5212048
BX537563.1 - 23, 24 5632

View Full Product Details