Gene Transcripts

Gene Symbol
CCDC85B
Entrez Gene ID
11007
Gene Name
coiled-coil domain containing 85B
Gene Aliases
DIPA
Chromosome Location
Chr. 11: 65890404 - 65891635 on Build GRCh38
UniGene ID
Hs.66713
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006848.2 NP_006839.2 1 1220
GenBank mRNA BC008796.2 - 1 985
U63825.1 - 1 868

View Full Product Details