Gene Transcripts

Gene Symbol
ZNF8
Entrez Gene ID
7554
Gene Name
zinc finger protein 8
Gene Aliases
HF.18, Zfp128
Chromosome Location
Chr. 19: 58278952 - 58295888 on Build GRCh38
UniGene ID
Hs.590941
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021089.2 NP_066575.2 4 652
GenBank mRNA AK093110.1 - 4 652
AK225825.1 - 4 649
BC039323.1 - 4 610 5267031

View Full Product Details