Gene Transcripts

Gene Symbol
OR10J3
Entrez Gene ID
441911
Gene Name
olfactory receptor family 10 subfamily J member 3
Gene Aliases
OR1-25, OR10J3P
Chromosome Location
Chr. 1: 159313670 - 159314659 on Build GRCh38
UniGene ID
Hs.553833
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004467.1 NP_001004467.1 1 151

View Full Product Details