Gene Transcripts

Gene Symbol
SLC39A2
Entrez Gene ID
29986
Gene Name
solute carrier family 39 member 2
Gene Aliases
6A1, ETI-1, ZIP-2, ZIP2
Chromosome Location
Chr. 14: 20999255 - 21001875 on Build GRCh38
UniGene ID
Hs.175783
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256588.1 NP_001243517.1 4 1261
NM_014579.3 NP_055394.2 4 1266
GenBank mRNA AF186081.1 - 4 1210
D83039.1 - 4 1204

View Full Product Details