Gene Transcripts

Gene Symbol
CHCHD5
Entrez Gene ID
84269
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 5
Gene Aliases
C2orf9, MIC14
Chromosome Location
Chr. 2: 112583606 - 112589040 on Build GRCh38
UniGene ID
Hs.375707
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001304353.1 NP_001291282.1 2 161
NM_001304354.1 NP_001291283.1 2 158
NM_032309.3 NP_115685.1 2 324
XM_005263824.3 XP_005263881.1 2 1155
GenBank mRNA AK096990.1 - 2 166
AK312219.1 - 2 300
BC004498.2 - 2 302 3831856
BM563093.1 - 2 158
BX115079.1 - 2 160
HY011716.1 - 2 319
N39730.1 - 2 160

View Full Product Details