Gene Transcripts

Gene Symbol
Sars
Entrez Gene ID
266975
Gene Name
seryl-tRNA synthetase
Gene Aliases
Sars1
Chromosome Location
Chr. 2: 211219743 - 211235475 on Build Rnor_6.0
UniGene ID
Rn.73067
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007606.1 NP_001007607.1 7 848
XM_006233136.2 XP_006233198.1 7 883
GenBank mRNA AY145052.1 - 3 275
BC061765.1 - 7 848 5620893

View Full Product Details