Gene Transcripts

Gene Symbol
Thyn1
Entrez Gene ID
300470
Gene Name
thymocyte nuclear protein 1
Gene Aliases
Chromosome Location
Chr. 8: 28045093 - 28053974 on Build Rnor_6.0
UniGene ID
Rn.87291
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007661.2 NP_001007662.1 3 347
GenBank mRNA BC064031.1 - 3 351
FQ213324.1 - 2 496

View Full Product Details