Gene Transcripts

Gene Symbol
Thoc6
Entrez Gene ID
386612
Gene Name
THO complex 6 homolog (Drosophila)
Gene Aliases
F830014G06Rik, Wdr58
Chromosome Location
Chr. 17: 23668618 - 23673903 on Build GRCm38
UniGene ID
Mm.328831
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008425.1 NP_001008425.1 13 1194
GenBank mRNA AK089746.1 - 10 1522
BC085131.1 - 13 1194 5715435
BC089551.1 - 14 1194 30303151

View Full Product Details