Gene Transcripts

Gene Symbol
CHAC2
Entrez Gene ID
494143
Gene Name
ChaC cation transport regulator homolog 2
Gene Aliases
Chromosome Location
Chr. 2: 53767792 - 53775150 on Build GRCh38
UniGene ID
Hs.585944
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008708.2 NP_001008708.1 3 581
GenBank mRNA BC017941.1 - 3 554
BC019239.1 - 3 585 4050858
BC025376.1 - 3 585 4045703
BC053896.1 - 3 558 6174351

View Full Product Details