Gene Transcripts

Gene Symbol
Vom1r19
Entrez Gene ID
494298
Gene Name
vomeronasal 1 receptor 19
Gene Aliases
V1rf6, V1rf7
Chromosome Location
Chr. 1: 61095171 - 61096112 on Build Rnor_6.0
UniGene ID
Rn.129450
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008954.1 NP_001008954.1 1 334
GenBank mRNA AY510367.1 - 1 334
AY510368.1 - 1 334

View Full Product Details