Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001003911.2 | NP_001003911.2 | 19 | 3860 | |
GenBank mRNA | AL359935.1 | - | 1 | 107 | |
AY551090.1 | - | 19 | 3719 | ||
BC058991.1 | - | 16 | 2832 | ||
BC076575.1 | - | 18 | 3568 | ||
BC141173.1 | - | 19 | 3636 | 9055785 | |
BC145491.1 | - | 19 | 3636 | 9054054 |