Gene Transcripts

Gene Symbol
PTDSS1
Entrez Gene ID
9791
Gene Name
phosphatidylserine synthase 1
Gene Aliases
LMHD, PSS1, PSSA
Chromosome Location
Chr. 8: 96261886 - 96334552 on Build GRCh38
UniGene ID
Hs.292579
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290225.1 NP_001277154.1 8 1085
NM_014754.2 NP_055569.1 10 1255
GenBank mRNA AK293036.1 - 10 1239
AK293513.1 - 6 565
BC002376.2 - 8 1058 2961075
BC004192.2 - 10 1205 3509418
BC004390.1 - 10 1205 3634879
BC004502.2 - 6 676
D14694.1 - 10 1202

View Full Product Details