Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001005535.2 | NP_001005535.1 | 2 | 573 | |
NM_001190820.1 | NP_001177749.1 | 2 | 573 | ||
GenBank mRNA | BC081871.1 | - | 2 | 527 | |
DY310785.1 | - | 2 | 597 | ||
DY317721.1 | - | 2 | 573 |