Gene Transcripts

Gene Symbol
Nol4
Entrez Gene ID
319211
Gene Name
nucleolar protein 4
Gene Aliases
1700013J13Rik, 4930568N03Rik, Gm1262
Chromosome Location
Chr. 18: 22693152 - 23041675 on Build GRCm38
UniGene ID
Mm.209896
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161483.1 NP_001154955.1 11 1923
NM_199024.2 NP_950189.2 10 3410
GenBank mRNA AK029800.1 - 6 1235
AK031008.1 - 10 3410
AK031753.1 - 4 916
AK081727.1 - 1 60
AK086574.1 - 7 1427
BC056377.1 - 11 1916 6850243

View Full Product Details