Gene Transcripts

Gene Symbol
ZBTB1
Entrez Gene ID
22890
Gene Name
zinc finger and BTB domain containing 1
Gene Aliases
ZNF909
Chromosome Location
Chr. 14: 64504574 - 64533690 on Build GRCh38
UniGene ID
Hs.605143
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001123329.1 NP_001116801.1 2 750
NM_014950.2 NP_055765.2 2 750
XM_005267410.1 XP_005267467.1 4 886
XM_011536566.2 XP_011534868.1 3 623
XM_011536568.2 XP_011534870.1 4 886
XM_017021094.1 XP_016876583.1 4 1762
XM_017021095.1 XP_016876584.1 3 678
GenBank mRNA AB023214.1 - 2 621
AK291743.1 - 2 528
AY598321.1 - 1 359
BC050719.1 - 4 790 6141266
BX248777.1 - 2 750

View Full Product Details