Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015356.4 | NP_056171.3 | 11 | 1242 | |
NM_182706.4 | NP_874365.3 | 11 | 1242 | ||
GenBank mRNA | AF240677.1 | - | 11 | 1235 | |
AF271734.2 | - | 11 | 1213 | ||
AY062238.1 | - | 11 | 1244 | ||
D63481.3 | - | 11 | 1237 |