Gene Transcripts

Gene Symbol
UBE2J1
Entrez Gene ID
51465
Gene Name
ubiquitin conjugating enzyme E2 J1
Gene Aliases
CGI-76, HSPC153, HSPC205, HSU93243, NCUBE-1, NCUBE1, UBC6, UBC6E, Ubc6p
Chromosome Location
Chr. 6: 89326625 - 89352917 on Build GRCh38
UniGene ID
Hs.163776
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016021.2 NP_057105.2 4 625
XM_011535887.1 XP_011534189.1 4 612
XM_011535888.2 XP_011534190.1 4 642
GenBank mRNA AF151039.1 - 1 18
AF151834.1 - 4 432
AF161502.1 - 4 320
AJ245898.1 - 4 356
AJ420578.1 - 3 274
AK223464.1 - 5 364
AK290574.1 - 4 467
AK300629.1 - 5 370
BC013973.1 - 4 587 4137664
BF058850.1 - 5 544
U93243.1 - 4 321

View Full Product Details