Gene Transcripts
- Gene Symbol
- WAPL
- Entrez Gene ID
- 23063
- Gene Name
- WAPL cohesin release factor
- Gene Aliases
- FOE, KIAA0261, WAPAL
- Chromosome Location
- Chr. 10: 86435256 - 86521816 on Build GRCh38
- UniGene ID
- Hs.203099
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001318328.1 |
NP_001305257.1
|
9, 10 |
2798 |
|
NM_015045.3 |
NP_055860.1
|
9, 10 |
2816 |
|
XM_011539548.1 |
XP_011537850.1
|
9, 10 |
2866 |
|
XM_017015979.1 |
XP_016871468.1
|
8, 9 |
2713 |
|
XM_017015980.1 |
XP_016871469.1
|
8, 9 |
2656 |
|
XM_017015981.1 |
XP_016871470.1
|
6, 7 |
2403 |
|
GenBank mRNA |
AB065003.1 |
- |
9, 10 |
2819 |
|
AF479418.1 |
- |
10, 11 |
2494 |
|
AK027005.1 |
- |
2, 3 |
539 |
|
BC017393.1 |
- |
2, 3 |
252 |
|
BC140858.1 |
- |
9, 10 |
2522 |
9021726 |
BC150269.1 |
- |
10, 11 |
2667 |
9007232 |
D87450.1 |
- |
10, 11 |
2667 |
|
View Full Product Details