Gene Transcripts

Gene Symbol
SOBP
Entrez Gene ID
55084
Gene Name
sine oculis binding protein homolog
Gene Aliases
JXC1, MRAMS
Chromosome Location
Chr. 6: 107490104 - 107661309 on Build GRCh38
UniGene ID
Hs.445244
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018013.3 NP_060483.3 6 1989
XM_005267041.4 XP_005267098.1 7 2150
XM_005267042.4 XP_005267099.1 6 2055
XM_011535920.2 XP_011534222.1 7 2150
XM_011535921.2 XP_011534223.1 7 1674
XM_011535923.2 XP_011534225.1 2 888
XM_017010991.1 XP_016866480.1 5 2512
XM_017010992.1 XP_016866481.1 5 1281
XM_017010993.1 XP_016866482.1 5 1311
XM_017010994.1 XP_016866483.1 5 1270
GenBank mRNA DQ507800.1 - 6 1485

View Full Product Details