Gene Transcripts

Gene Symbol
TBC1D31
Entrez Gene ID
93594
Gene Name
TBC1 domain family member 31
Gene Aliases
Gm85, WDR67
Chromosome Location
Chr. 8: 123041961 - 123165169 on Build GRCh38
UniGene ID
Hs.492716
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145088.1 NP_001138560.1 7 1024
NM_145647.3 NP_663622.2 7 1024
XM_005251102.1 XP_005251159.1 7 1024
XM_005251103.1 XP_005251160.1 7 1024
XM_005251104.1 XP_005251161.1 7 1024
XM_011517378.2 XP_011515680.1 7 1539
XM_011517379.2 XP_011515681.1 7 1024
XM_017013983.1 XP_016869472.1 7 1024
XM_017013984.1 XP_016869473.1 6 1146
XM_017013985.1 XP_016869474.1 6 1019
XM_017013986.1 XP_016869475.1 6 880
XM_017013987.1 XP_016869476.1 7 1024
XM_017013988.1 XP_016869477.1 6 859
XM_017013989.1 XP_016869478.1 6 1022
XM_017013990.1 XP_016869479.1 6 822
GenBank mRNA AK056434.1 - 7 1025
AK094612.1 - 6 822
BC026969.1 - 3 345
BC101724.1 - 7 991 8069230
BC105800.1 - 7 985
BC111954.1 - 7 991 8327422
BC143523.1 - 6 875
BC143525.1 - 7 991 9052034

View Full Product Details