Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001032409.2 | NP_001027581.1 | 1 | 397 | |
NM_001320151.1 | NP_001307080.1 | 1 | 397 | ||
NM_002534.3 | NP_002525.2 | 1 | 397 | ||
NM_016816.3 | NP_058132.2 | 1 | 397 | ||
XM_006719434.1 | XP_006719497.1 | 1 | 224 | ||
XM_011538413.1 | XP_011536715.1 | 1 | 224 | ||
XM_017019361.1 | XP_016874850.1 | 1 | 224 | ||
XM_017019362.1 | XP_016874851.1 | 1 | 96 | ||
GenBank mRNA | AJ629455.1 | - | 1 | 167 | |
AK223006.1 | - | 1 | 156 | ||
AK225384.1 | - | 1 | 197 | ||
AK291003.1 | - | 1 | 201 | ||
AK300346.1 | - | 1 | 122 | ||
AK301498.1 | - | 1 | 156 | ||
AY730627.1 | - | 1 | 143 | ||
AY730628.1 | - | 1 | 143 | ||
BC000562.2 | - | 1 | 204 | 3162421 | |
BC061587.1 | - | 1 | 165 | 6502031 | |
BC071981.1 | - | 1 | 200 | 4053409 | |
BT006785.1 | - | 1 | 134 | ||
CB112445.1 | - | 1 | 397 | ||
D00068.1 | - | 1 | 197 | ||
X02874.1 | - | 1 | 167 | ||
X04371.1 | - | 1 | 227 |