Gene Transcripts
- Gene Symbol
- RAB6B
- Entrez Gene ID
- 51560
- Gene Name
- RAB6B, member RAS oncogene family
- Gene Aliases
- Chromosome Location
- Chr. 3: 133824235 - 133895847 on Build GRCh38
- UniGene ID
- Hs.715344
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_016577.3 |
NP_057661.3
|
2, 3 |
518 |
|
XM_011512893.2 |
XP_011511195.1
|
2, 3 |
193 |
|
GenBank mRNA |
AF091031.1 |
- |
1, 2 |
65 |
|
AF166492.1 |
- |
2, 3 |
474 |
|
AF498940.1 |
- |
2, 3 |
137 |
|
AK295451.1 |
- |
3, 4 |
255 |
|
AK312378.1 |
- |
2, 3 |
438 |
|
BC002510.2 |
- |
2, 3 |
487 |
3050592 |
BC078662.1 |
- |
2, 3 |
266 |
6450986 |
BT007263.1 |
- |
2, 3 |
137 |
|
View Full Product Details