Gene Transcripts

Gene Symbol
Cirh1a
Entrez Gene ID
291987
Gene Name
cirrhosis, autosomal recessive 1A (cirhin)
Gene Aliases
Cirhin, Naic, Teg-292, Tex292
Chromosome Location
Chr. 19: 39087995 - 39116124 on Build Rnor_6.0
UniGene ID
Rn.106235
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009640.1 NP_001009640.1 14, 15 1686
XM_006255562.1 XP_006255624.1 14, 15 1732
XM_006255563.2 XP_006255625.1 14, 15 1895
GenBank mRNA BC088461.1 - 14, 15 1686

View Full Product Details