Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001009663.1 | NP_001009663.1 | 2 | 143 | |
GenBank mRNA | BC088300.1 | - | 2 | 143 | |
FQ209415.1 | - | 2 | 188 | ||
FQ211289.1 | - | 2 | 158 | ||
FQ218294.1 | - | 2 | 163 | ||
FQ218398.1 | - | 2 | 152 | ||
FQ219289.1 | - | 2 | 217 |