Gene Transcripts

Gene Symbol
Ribc2
Entrez Gene ID
300122
Gene Name
RIB43A domain with coiled-coils 2
Gene Aliases
RGD1309033
Chromosome Location
Chr. 7: 126028371 - 126043371 on Build Rnor_6.0
UniGene ID
Rn.96045
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013949.1 NP_001013971.2 3 447
GenBank mRNA BC079434.1 - 3 447

View Full Product Details