Gene Transcripts

Gene Symbol
Slc10a5
Entrez Gene ID
310230
Gene Name
solute carrier family 10, member 5
Gene Aliases
Chromosome Location
Chr. 2: 93669962 - 93671266 on Build Rnor_6.0
UniGene ID
Rn.162564
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025280.1 NP_001020451.1 1 868
GenBank mRNA DQ074435.1 - 1 868

View Full Product Details