Gene Transcripts

Gene Symbol
Prm1
Entrez Gene ID
24685
Gene Name
protamine 1
Gene Aliases
Chromosome Location
Chr. 10: 4945995 - 4946241 on Build Rnor_6.0
UniGene ID
Rn.27657
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002850.1 NP_001002850.1 2 138
GenBank mRNA BC126070.1 - 2 222

View Full Product Details