Gene Transcripts

Gene Symbol
OR2G6
Entrez Gene ID
391211
Gene Name
olfactory receptor family 2 subfamily G member 6
Gene Aliases
Chromosome Location
Chr. 1: 248521647 - 248522597 on Build GRCh38
UniGene ID
Hs.553784
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013355.1 NP_001013373.1 1 814
GenBank mRNA BC137245.1 - 1 846
BC137266.1 - 1 846 9021263

View Full Product Details