Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001014152.1 | NP_001014174.1 | 17 | 1645 | |
XM_006255269.2 | XP_006255331.1 | 17 | 1682 | ||
XM_006255270.2 | XP_006255332.1 | 17 | 1682 | ||
XM_008772410.1 | XP_008770632.1 | 17 | 2394 | ||
GenBank mRNA | BC085909.1 | - | 17 | 1645 |