Gene Transcripts

Gene Symbol
Zfp52
Entrez Gene ID
361487
Gene Name
zinc finger protein 52
Gene Aliases
Chromosome Location
Chr. 1: 61388812 - 61509202 on Build Rnor_6.0
UniGene ID
Rn.99395
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014158.1 NP_001014180.1 6 1238
XM_006228003.2 XP_006228065.1 5 971
GenBank mRNA BC085827.1 - 6 1238

View Full Product Details