Gene Transcripts

Gene Symbol
P2RY11
Entrez Gene ID
5032
Gene Name
purinergic receptor P2Y11
Gene Aliases
P2Y11
Chromosome Location
Chr. 19: 10111521 - 10115389 on Build GRCh38
UniGene ID
Hs.14468
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002566.4 NP_002557.2 2 1331
GenBank mRNA BC073827.1 - 2 1314

Gene Symbol
PPAN-P2RY11
Entrez Gene ID
692312
Gene Name
PPAN-P2RY11 readthrough
Gene Aliases
Chromosome Location
Chr. 19: 10106223 - 10115389 on Build GRCh38
UniGene ID
Hs.14468
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040664.2 NP_001035754.1 13 2562
NM_001198690.1 NP_001185619.1 13 2624
GenBank mRNA BC171852.1 - 13 2624

View Full Product Details