Gene Transcripts

Gene Symbol
Psg19
Entrez Gene ID
26439
Gene Name
pregnancy specific glycoprotein 19
Gene Aliases
CGM7, Cea-4, Cea4
Chromosome Location
Chr. 7: 18789125 - 18798542 on Build GRCm38
UniGene ID
Mm.36878
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011964.2 NP_036094.2 5 1766
GenBank mRNA AA408180.1 - 1 117
BC099467.1 - 5 1742 30713077
X98111.1 - 5 1766
X98112.1 - 4 1406

View Full Product Details