Gene Transcripts

Gene Symbol
VPS8
Entrez Gene ID
23355
Gene Name
VPS8, CORVET complex subunit
Gene Aliases
KIAA0804
Chromosome Location
Chr. 3: 184812137 - 185052614 on Build GRCh38
UniGene ID
Hs.269263
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009921.2 NP_001009921.1 16 1416
NM_015303.3 NP_056118.2 15 1410
XM_005247251.3 XP_005247308.1 16 1684
XM_005247253.4 XP_005247310.1 16 1422
XM_006713556.3 XP_006713619.1 8 704
XM_011512599.2 XP_011510901.1 3 166
XM_011512600.2 XP_011510902.1 8 618
XM_011512601.2 XP_011510903.1 8 704
XM_017006040.1 XP_016861529.1 14 1104
XM_017006041.1 XP_016861530.1 16 1422
GenBank mRNA AB018347.1 - 8 595
AK056661.1 - 16 1388
AK292423.1 - 15 1382
AL833838.1 - 13 949
BC140768.1 - 16 1352 9021636
BX647915.1 - 15 1371

View Full Product Details