Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001010854.1 | NP_001010854.1 | 7 | 1038 | |
NM_001320421.1 | NP_001307350.1 | 7 | 1034 | ||
XM_005267367.1 | XP_005267424.1 | 3 | 349 | ||
XM_011536497.1 | XP_011534799.1 | 7 | 1096 | ||
XM_017021044.1 | XP_016876533.1 | 7 | 1002 | ||
XM_017021045.1 | XP_016876534.1 | 7 | 1002 | ||
XM_017021046.1 | XP_016876535.1 | 7 | 1096 | ||
XM_017021047.1 | XP_016876536.1 | 6 | 713 | ||
GenBank mRNA | AL832848.1 | - | 6 | 716 | |
BC035865.2 | - | 1 | 139 | ||
BC048270.2 | - | 1 | 139 | ||
BX248275.1 | - | 3 | 330 |