Gene Transcripts

Gene Symbol
Sh2d4a
Entrez Gene ID
306376
Gene Name
SH2 domain containing 4A
Gene Aliases
Chromosome Location
Chr. 16: 23084247 - 23156825 on Build Rnor_6.0
UniGene ID
Rn.137349
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012048.1 NP_001012048.1 9 1906
XM_006253032.2 XP_006253094.1 9 1919
GenBank mRNA BC079100.1 - 9 1906

View Full Product Details