Gene Transcripts

Gene Symbol
Tst
Entrez Gene ID
25274
Gene Name
thiosulfate sulfurtransferase
Gene Aliases
RHODAN
Chromosome Location
Chr. 7: 119616323 - 119623072 on Build Rnor_6.0
UniGene ID
Rn.6360
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012808.1 NP_036940.1 1 300
GenBank mRNA BC088449.1 - 1 300
X56228.1 - 1 263

View Full Product Details