Gene Transcripts
- Gene Symbol
- STXBP4
- Entrez Gene ID
- 252983
- Gene Name
- syntaxin binding protein 4
- Gene Aliases
- Synip
- Chromosome Location
- Chr. 17: 54968728 - 55164285 on Build GRCh38
- UniGene ID
- Hs.35199
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_178509.5 |
NP_848604.3
|
4, 5 |
395 |
|
XM_005257187.4 |
XP_005257244.1
|
4, 5 |
431 |
|
XM_006721797.3 |
XP_006721860.1
|
4, 5 |
429 |
|
XM_006721798.3 |
XP_006721861.1
|
4, 5 |
426 |
|
XM_017024410.1 |
XP_016879899.1
|
4, 5 |
432 |
|
XM_017024411.1 |
XP_016879900.1
|
4, 5 |
430 |
|
XM_017024412.1 |
XP_016879901.1
|
4, 5 |
429 |
|
XM_017024414.1 |
XP_016879903.1
|
4, 5 |
431 |
|
XM_017024415.1 |
XP_016879904.1
|
4, 5 |
431 |
|
GenBank mRNA |
AK122865.1 |
- |
4, 5 |
395 |
|
BC125116.1 |
- |
4, 5 |
384 |
40118892 |
View Full Product Details