Gene Transcripts

Gene Symbol
FREM3
Entrez Gene ID
166752
Gene Name
FRAS1 related extracellular matrix 3
Gene Aliases
Chromosome Location
Chr. 4: 143577407 - 143700675 on Build GRCh38
UniGene ID
Hs.252714
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001168235.1 NP_001161707.1 5 5684
GenBank mRNA DN831271.1 - 3 323

View Full Product Details