Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018256.3 | NP_060726.3 | 2, 3 | 884 | |
XM_011511469.2 | XP_011509771.1 | 2, 3 | 877 | ||
GenBank mRNA | AF242546.1 | - | 2, 3 | 267 | |
AK001743.1 | - | 2, 3 | 321 | ||
AK022781.1 | - | 2, 3 | 329 | ||
AK022782.1 | - | 2, 3 | 329 | ||
AK056092.1 | - | 2, 3 | 884 | ||
BC008082.2 | - | 2, 3 | 264 | 3050219 | |
EF011617.1 | - | 2, 3 | 263 |