Gene Transcripts

Gene Symbol
CWF19L1
Entrez Gene ID
55280
Gene Name
CWF19-like 1, cell cycle control (S. pombe)
Gene Aliases
C19L1, SCAR17, hDrn1
Chromosome Location
Chr. 10: 100232296 - 100267681 on Build GRCh38
UniGene ID
Hs.215502
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001303404.1 NP_001290333.1 4 342
NM_001303405.1 NP_001290334.1 4 387
NM_001303407.1 NP_001290336.1 4 326
NM_018294.5 NP_060764.3 4 342
GenBank mRNA AK001860.1 - 4 288
AK097895.1 - 4 325
AK225590.1 - 4 300
AL832515.1 - 4 299
BC008746.2 - 4 283 3345869
DC299678.1 - 4 342
KU178692.1 - 4 254
KU178693.1 - 4 238

View Full Product Details