Gene Transcripts

Gene Symbol
LAPTM4B
Entrez Gene ID
55353
Gene Name
lysosomal protein transmembrane 4 beta
Gene Aliases
LAPTM4beta, LC27
Chromosome Location
Chr. 8: 97775581 - 97852602 on Build GRCh38
UniGene ID
Hs.492314
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018407.4 NP_060877.3 7 1546
GenBank mRNA AK075326.1 - 7 1308
AK311377.1 - 2 785
AL136942.1 - 7 1300
AY057051.2 - 7 1546
AY261384.1 - 7 1565
BC014129.2 - 7 1302 4581284
BC031021.1 - 7 1327 4690368
BC038117.1 - 7 2094 5264567

View Full Product Details