Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001304506.1 | NP_001291435.1 | 2 | 149 | |
NM_019110.4 | NP_061983.2 | 2 | 814 | ||
XM_005249095.3 | XP_005249152.1 | 4 | 858 | ||
XM_005249097.3 | XP_005249154.1 | 4 | 554 | ||
XM_005249098.3 | XP_005249155.1 | 2 | 472 | ||
XM_005249100.4 | XP_005249157.2 | 2 | 205 | ||
XM_006715095.3 | XP_006715158.1 | 3 | 331 | ||
XM_011514588.2 | XP_011512890.1 | 2 | 714 | ||
XM_011514589.2 | XP_011512891.1 | 2 | 270 | ||
XM_017010844.1 | XP_016866333.1 | 4 | 1509 | ||
GenBank mRNA | AK302017.1 | - | 2 | 130 | |
AK315783.1 | - | 2 | 784 | ||
AY781778.1 | - | 2 | 784 | ||
BC014031.2 | - | 2 | 742 | 4336904 | |
KU178905.1 | - | 2 | 498 | ||
KU178906.1 | - | 2 | 511 |