Gene Transcripts

Gene Symbol
UNCX
Entrez Gene ID
340260
Gene Name
UNC homeobox
Gene Aliases
UNCX4.1
Chromosome Location
Chr. 7: 1233018 - 1236977 on Build GRCh38
UniGene ID
Hs.232272
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080461.1 NP_001073930.1 3 596
GenBank mRNA KM587717.1 - 3 1232

View Full Product Details